Learn More
Lucifer Yellow CH Dipotassium Salt (Technical Grade), TRC
Product Code. 30179158
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179158 25 mg 25mg
30178917 50 mg 50mg
30179453 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30179158 Supplier Toronto Research Chemicals Supplier No. TRCL47372325MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 71206-95-6
Molecular Formula C13H9K2N5O9S2
Molecular Weight (g/mol) 521.57
Synonym 6-Amino-2-[(hydrazinocarbonyl)amino]-2,3-dihydro-1,3-dioxo-1H-benz[de]isoquinoline-5,8-disulfonic Acid Dipotassium Salt
SMILES O=[S](OC1=CC(C)=NN1C2=CC=CC=C2)(O)=O
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C13H11N5O9S2.2K/c14-10-5-1-4(28(22,23)24)2-6-9(5)7(3-8(10)29(25,26)27)12(20)18(11(6)19)17-13(21)16-15;;/h1-3H,14-15H2,(H2,16,17,21)(H,22,23,24)(H,25,26,27);;/q;2*+1/p-2
Chemical Name or Material Lucifer Yellow CH Dipotassium Salt