Learn More
Lubricating Oil Standard 11: Ca @ 0.050; P @ 0.150; S @ 0.200; Zn @ 0.200 wt% in Lubricating Oil, VHG™
Description
Lubricating Oil Standard 11: Ca @ 0.050; P @ 0.150; S @ 0.200; Zn @ 0.200 wt% in Lubricating Oil
Specifications
Specifications
| Chemical Name or Material | Calcium/Phosphorus/Sulfur/Zinc |
| CAS | 7440-70-2, 7723-14-0, 7704-34-9, 7440-66-6 |
| Methods | XRF |
| Analyte or Component Names | Calcium/Phosphorus/Sulfur/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | 20 cSt Mineral Oil |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Calcium 0.0501%/Phosphorus 0.1504%/Sulfur 0.1999%/Zinc 0.2001% |