Learn More
IKA™ LR 2000.2 Reactor Vessel
Product Code. 10498671 Shop All IKA Products
Change view
Click to view available options
For Use With (Equipment):
For LR-2.ST and LR 2000 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10498671 For LR-2.ST and LR 2000 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10498671 Supplier IKA™ Supplier No. 0002509600
Only null left
Add to basket
Add to basket
Use this vessel for the optimization and reproduction of various chemical reactions, mixing and homogenization processes. IKA™ LR 2000.1 Reactor Vessel features double walled glass, quick-action connectors and bottom discharge valve.
  • For LR-2.ST and LR 2000 V systems
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) For LR-2.ST and LR 2000 V
Product Type Reactor Vessel