Learn More
Miele Professional™ Lower Basket A 150
Product Code. 15382579 Shop All Miele Products
Change view
Click to view available options
Color:
Stainless Steel
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15382579 Stainless Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15382579 Supplier Miele Professional™ Supplier No. 12666330
  • ONLINE EXCLUSIVE PRICE
    847.00 EUR
    557.00 EUR / Pack of 1
    Save 290.00 EUR
    34% Off
Estimated Shipment: 27-11-2026
Lower basket A 150 for the optimum loading of injector modules or inserts
  • Lower basket with two module connections
  • for holding up to 2 injector modules or inserts
  • Automatic closing of the connections when not in use
  • Clearances: width 494, length 440 mm
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel
Product Type Basket
Color Stainless Steel
Rack Type Lower basket for modules
For Use With (Equipment) PG 8583/PG8593 laboratory washers
Includes Two docking pipes, self-sealing docking valves
Depth (Metric) 546 mm
Height (Metric) 154 mm
Width (Metric) 529 mm