Learn More
Lenz Laborglasinstrumente™ Low Shape Weighing Bottles
Product Code. 11826174
Change view
Click to view available options
Capacity (Metric):
10 mL
15 mL
45 mL
Unit Size:
Each
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11826174 45 mL Each
11758104 10 mL Pack of 6
11768104 15 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11826174 Supplier Lenz Laborglasinstrumente™ Supplier No. 05565380
  • ONLINE EXCLUSIVE PRICE
    66.00 EUR
    48.83 EUR / Pack of 1
    Save 17.17 EUR
    26% Off
Estimated Shipment: 16-12-2026
Specifications
Capacity (Metric) 45 mL
Includes Interchangeable NS stopper, inscription label
Quantity 1Pack
Stopper Number NS 80/12
Product Line Bottle
Product Type Weighing Bottle