Learn More
Scotch-Brite™ Low Scratch Scour Sponge, 3001HEX
Product Code. 17158860
Change view
Click to view available options
Dimensions (L x W):
113 x 98 mm
Unit Size:
Pack of 16
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
17158860 113 x 98 mm Pack of 16
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17158860 Supplier Scotch-Brite™ Supplier No. 7100202994
  • ONLINE EXCLUSIVE PRICE
    73.00 EUR
    22.96 EUR / Pack of 16
    Save 50.04 EUR
    68% Off
Estimated Shipment: 10-12-2026
Two-in-one, low scratch cleaning sponge designed for cleaning delicate surfaces. Features a raised power dot side that is engineered to scour away tough messes like a medium duty pad, but with less visible scratches. Includes a sponge to wipe spills and messes.
  • Designed to clean a wide variety of surfaces where minimal scratching is desired
  • Raised power dot side is engineered to prevent large food particles from getting trapped for fast cleanup
  • Ergnomic hexagon shape fits comfortably in the hand
  • Easily cleans hard to reach areas
  • HACCP certified
TRUSTED_SUSTAINABILITY
Specifications
Product Type Scouring Pad/Sponge
Dimensions (L x W) 113 x 98 mm
For Use With (Application) Cleaning Delicate Surfaces
Length (Metric) 113 mm
Model 3001 HEX
Quantity 16 Pk.
Style 2 in. 1
Width (Metric) 98 mm