Learn More
Thermo Scientific Chemicals Low Form Inconel Crucible;Cap (ml), 500;Top Dia (mm), 102;Bottom Dia (mm), 89;Depth (mm), 66
Product Code. 15423585
Change view
Click to view available options
Capacity (Metric):
15 mL
25 mL
55 mL
75 mL
100 mL
250 mL
500 mL
Diameter (Metric) Outer:
33mm
45mm
46mm
47mm
51mm
59mm
82mm
102mm
Height (Metric):
23mm
35mm
43mm
46mm
60mm
66mm
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Outer Height (Metric) unitSize
15423585 15 mL 33mm 23mm Each
15453595 25 mL 45mm 23mm Each
15473605 55 mL 46mm 35mm Each
15413615 55 mL 47mm 43mm Each
15473615 75 mL 51mm 43mm Each
15453625 500 mL 102mm 66mm Each
15403635 100 mL 59mm 46mm Each
15453635 250 mL 82mm 60mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15423585 Supplier Thermo Scientific Chemicals Supplier No. 035886.KT
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Description Low Form
Material Inconel
Capacity (Metric) 15 mL
Diameter (Metric) Outer 33mm
Height (Metric) 23mm
Product Type Inconel Crucible
Diameter (Metric) Base 25mm