Learn More
Lovibond™ DPD No. 1 HR Tablets
Click to view available options
Range:
0.01 to 10ppm
Unit Size:
100 tablets
Description
Determine the amount of free chlorine in your hot tub, swimming pool, or spa water with these foil-packaged, fast-acting tablets. Lovibond™ DPD No. 1 HR Tablets are foil-packaged tablets for consistent and reliable reagent delivery.
- Simple, time-saving and practical
- Foil packaged test tablets remain the most consistent and reliable reagent delivery system available
- Each tablet is hermetically sealed within an individually formed foil cup
Specifications
Specifications
| Color | White |
| Composition | N,N-diethylbenzene-1,4-diammonium Sulphate 2.5 to 5%, Citric Acid 0.1 to 1%, non-hazardous components 20 to 35%, Sodium cyclamate 60 to 70%, Polyvinyl Alcohol ≤0.1%, Di-Sodium Dihydrogenethylenediaminetetraacetate ≤2.5% |
| pH Range | 6.3 |
| Format | Tablet |
| For Use With (Application) | Free chlorine analysis in water |
| For Use With (Equipment) | SC450 chlorine 2 and 3 colorimeters, MC500 colorimeter, SP600 spectrophotometer, AquaPRO3 and PRO6 Pool photometers |
| Range | 0.01 to 10ppm |
| Storage Requirements | 20°C |
| Test Method | DPD |
| Type | DPD No. 1 HR Tablet |