missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LOR Polyclonal Antibody

Description
Recommended positive controls: Raji, HeLa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes loricrin, a major protein component of the cornified cell envelope found in terminally differentiated epidermal cells. Mutations in this gene are associated with Vohwinkel's syndrome and progressive symmetric erythrokeratoderma, both inherited skin diseases.
Specifications
Specifications
| Antigen | LOR |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.28 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | LORICRIN |
| Gene Accession No. | P18165, P23490 |
| Gene Alias | AI036317; LOR; Loricrin; loricrin cornified envelope precursor protein; LRN; MGC111513; RGD1559993; S77319 |
| Gene Symbols | LORICRIN |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?