missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lopinavir, MedChemExpress
Description
Lopinavir (ABT-378) is a highly potent, selective peptidomimetic inhibitor of the HIV-1 protease, with Kis of 1.3 to 3.6 pM for wild-type and mutant HIV protease. Lopinavir acts by arresting maturation of HIV-1 thereby blocking its infectivity. Lopinavir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 14.2 μM.
Specifications
Specifications
| Chemical Name or Material | Lopinavir |
| Physical Form | Solid |
| CAS | 192725-17-0 |
| Color | White |
| Quantity | 250 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C37H48N4O5 |
| For Use With (Application) | COVID-19-anti-virus |
| Synonym | ABT-378 |
| Solubility Information | DMSO : 250 mg/mL (397.58 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?