Learn More
Lonza™ DNA QuantLadders
Description
• Ladders are prepared from five highly purified DNA fragments in perfect relative mass equivalence
• Reverse QuantLadders reverse the mass equivalence order of the QuantLadder
• SimplyLoad™ ready-to-load ladders contain loading buffer; other ladders are supplied with separate loading buffer
• Store at 2° to 8°C for up to 12 months
Specifications
Specifications
| Buffer | TE |
| Concentration | 100 μg/mL |
| Form | Liquid |
| Formulation | Includes a vial of 6X sample loading buffer 1mL total volume 15% w/v Ficoll™ 400, 0.06% w/v Bromophenol Blue, 0.06% w/v Xylene Cyanol FF 30mM EDTA |
| For Use With (Application) | Quantitation of DNA within ±1% accuracy |
| No. of Bands | 5 |
| Quantity | 125 μL |
| Storage Requirements | 2° to 8°C |
| Product Type | DNA QuantLadder |