Learn More
Traceable™ Long Stem Digital Pocket Thermometer with Calibration
Product Code. 15412761 Shop All Cole Parmer Products
Change view
Click to view available options
Temperature Range (Metric):
-50°C to 300°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Unit Size
15412761 -50°C to 300°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15412761 Supplier Traceable™ Supplier No. 9446040
  • ONLINE EXCLUSIVE PRICE
    49.05 EUR
    38.43 EUR / Pack of 1
    Save 10.62 EUR
    21% Off
View all promotions
Estimated Shipment: 11-12-2026
These thermometers are ideal for laboratory, field, and process measurements.
  • Get quick temperature readings with guaranteed accuracy
  • Includes NIST-Traceable certificate
  • Long stainless steel stem is perfect for measurements in larger containers
  • Digital indicator
  • Pen-style design
Specifications
Certifications/Compliance NIST Traceable certificate supplied, ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA
Display Dimensions 0.3 in.
Includes 1.5 V battery
Length (English) Probe 11.5 in.
Probe Type Stainless steel
Resolution 0.1°; 1° above 199.9° and below -19°
Product Type Digital Indicator
Description Pen-style digital pocket thermometer
Max. Temperature (Metric) 300°C
Temperature Range (Metric) -50°C to 300°C
Battery Type 1.5 V
Accuracy ±1.0°C
Display LCD