missing translation for 'onlineSavingsMsg'
Learn More

Lomefloxacin, TRC

Product Code. 30137388
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137388 100 mg 100mg
30133932 50 mg 50mg
30007592 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137388 Supplier Toronto Research Chemicals Supplier No. TRCL469418100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98079-51-7
Molecular Formula C17 H19 F2 N3 O3
Molecular Weight (g/mol) 351.3479
Synonym 3-Quinolinecarboxylic acid, 1-ethyl-6,8-difluoro-1,4-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-, 1-Ethyl-6,8-difluoro-1,4-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid, 1-Ethyl-6,8-difluoro-7-(3-methyl-1-piperazinyl)-4-oxo-1,4-dihydro-3-quinoline carboxylic acid, DM 10, DM 10 (bactericide), Lomefloxacin, SC 47111A
IUPAC Name 1-ethyl-6,8-difluoro-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid
SMILES CCN1C=C(C(=O)O)C(=O)c2cc(F)c(N3CCNC(C)C3)c(F)c12

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H19F2N3O3/c1-3-21-8-11(17(24)25)16(23)10-6-12(18)15(13(19)14(10)21)22-5-4-20-9(2)7-22/h6,8-9,20H,3-5,7H2,1-2H3,(H,24,25)
Formula Weight 351.1394
Percent Purity >95%
Chemical Name or Material Lomefloxacin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.