Learn More
Notrax Logo Standard™ Affordable Absorbent Printed Logo Mat
Product Code. 17762018
Change view
Click to view available options
Dimensions (L x W):
90 x 60 cm
120 x 90 cm
150 x 90 cm
300 x 90 cm
180 x 120 cm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
17762018 300 x 90 cm Each
17722018 90 x 60 cm Each
17732018 120 x 90 cm Each
17742018 150 x 90 cm Each
17752018 180 x 120 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17762018 Supplier Notrax Supplier No. 193S0310WH
  • ONLINE EXCLUSIVE PRICE
    284.04 EUR
    230.97 EUR / Pack of 1
    Save 53.07 EUR
    18% Off
Estimated Shipment: 10-12-2026
Logo Standard™ can be fully customized with BrilliantStep™ state-of-the-art printing technology allowing 3-dimensional effects, fine lines and color shading, for each mat to have its truly individual image.
  • Logo Standard™ is the ideal promotional matting solution for light duty applications
  • Using BrilliantStep™ full color printing with almost infinite color combinations, the even velour surface allows fine printing and true color presentation
  • A durable vinyl backing with smooth finish adapts to any surface
  • Logo Standard™ can be fully customized with BrilliantStep™ state-of-the-art printing technology allowing 3-dimensional effects, fine lines and color shading, for each mat to have its truly individual image
Specifications
Dimensions (L x W) 300 x 90 cm
Style Frieze
Color Logo
Length (English) 118 in.
Length (Metric) 300 cm
Material Polyamide 6.6 High Twist Yarns
Type Industrial and Commercial Matting
Width (Metric) 90 cm