Learn More
Testo™ ComSoft Basic Software
Click to view available options
For Use With (Equipment):
ComSoft Basic
Unit Size:
Each
Description
- Software for programming and readout of Testo data loggers
- Free download of the ComSoft Basic (with registration) at www.testo.com
- Offers all basic functions of a logger software
- Runs on Windows XP ServicePack 3 (SP3), Windows Vista, Windows 7 and more
- Intuitive user interface guides the user through the individual processes step by step
- Graphic measurement value curve for clear presentation of the measurement values
- Logger configuration: start and stop criterion, measurement interval, storage rate and limit value adjustment
- Convenient export functions e.g. for the further processing of the data in Microsoft Excel, or the generation of a PDF which can be made available to other users
Specifications
Specifications
| Type | Datalogger Software |
| For Use With (Application) | Programming and readout of Testo data loggers, graphic and tabular measurement value presentation as well as export function |
| For Use With (Equipment) | ComSoft Basic |