Learn More
- Home
- Products
- Thermometers and Temperature Measurement
- Manual Thermometers
- Brannan™ LO-tox™ Lab Thermometer with 3-points UKAS Calibration
Brannan™ LO-tox™ Lab Thermometer with 3-points UKAS Calibration
Product Code. 17843298
Change view
Click to view available options
Temperature Range (Metric):
-1°C to 101°C
-1°C to 51°C
-10°C to +110°C
-10°C to +210°C
-10°C to +260°C
-10°C to +300°C
-10°C to +360°C
-10°C to +50°C
-20°C to +110°C
-20°C to +150°C
-35°C to +50°C
Subdivision:
0.1°C
0.2°C
0.5°C
1°C
2°C
Immersion Type:
Partial Immersion
Total Immersion
Length (Metric):
15.5 cm
30.5 cm
40.5 cm
Unit Size:
Each
24 product options available for selection
| Product Code. | Temperature Range (Metric) | Subdivision | Immersion Type | Length (Metric) | Unit Size |
|---|---|---|---|---|---|
| 17843298 | -1°C to 101°C | 0.2°C | Partial Immersion | 40.5 cm | Each |
| 17882988 | -1°C to 101°C | 0.2°C | Total Immersion | 40.5 cm | Each |
| 17892978 | -1°C to 51°C | 0.1°C | Partial Immersion | 40.5 cm | Each |
| 17872988 | -10°C to +360°C | 2°C | Partial Immersion | 30.5 cm | Each |
| 17832988 | -1°C to 51°C | 0.2°C | Partial Immersion | 40.5 cm | Each |
| 17802998 | -20°C to +150°C | 1°C | Partial Immersion | 30.5 cm | Each |
| 17893288 | -10°C to +260°C | 1°C | Partial Immersion | 40.5 cm | Each |
| 17833008 | -10°C to +110°C | 1°C | Total Immersion | 15.5 cm | Each |
| 17853068 | -20°C to +110°C | 0.5°C | Total Immersion | 30.5 cm | Each |
| 17843068 | -10°C to +210°C | 1°C | Partial Immersion | 40.5 cm | Each |
| 17803078 | -20°C to +110°C | 1°C | Partial Immersion | 30.5 cm | Each |
| 17863288 | -20°C to +110°C | 1°C | Total Immersion | 30.5 cm | Each |
| 17803298 | -20°C to +150°C | 1°C | Total Immersion | 30.5 cm | Each |
| 17893068 | -10°C to +360°C | 2°C | Total Immersion | 30.5 cm | Each |
| 17813118 | -35°C to +50°C | 1°C | Total Immersion | 30.5 cm | Each |
| 17833088 | -10°C to +210°C | 1°C | Total Immersion | 40.5 cm | Each |
| 17833068 | -1°C to 51°C | 0.2°C | Total Immersion | 40.5 cm | Each |
| 17813078 | -20°C to +110°C | 0.5°C | Partial Immersion | 30.5 cm | Each |
| 17813298 | -35°C to +50°C | 0.5°C | Partial Immersion | 30.5 cm | Each |
| 17853298 | -10°C to +50°C | 0.5°C | Partial Immersion | 30.5 cm | Each |
| 17883108 | -10°C to +50°C | 0.5°C | Total Immersion | 30.5 cm | Each |
| 17883298 | -1°C to 51°C | 0.1°C | Total Immersion | 40.5 cm | Each |
| 17873068 | -10°C to +300°C | 1°C | Partial Immersion | 30.5 cm | Each |
| 17843078 | -10°C to +50°C | 1°C | Total Immersion | 15.5 cm | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
This item is not returnable.
View return policy
Product Code.
17843298
Supplier
Brannan™
Supplier No.
44/824/0 + 49/503/0
-
162.00 EUR
137.40 EUR / Pack of 1
Save 24.60 EUR
15% Off
You must Sign In and have a business account to purchase this item.
Restricted Item
Estimated Shipment: 19-01-2027
Log in to see stock.
Log in to see stock.
Add to basket
123.66 EUR valid until 2026-11-30
BEST PRICE on Promo! Use promo code "27997" to get your promotional price.
BEST PRICE on Promo! Use promo code "27997" to get your promotional price.
Add to basket
This item is not returnable.
View return policy
Description
- LO-tox™ is an organic filling specially chosen to achieve a wide temperature span and provide the lowest practical levels of hazard
- Safe non-mercury filling
- Supplied with calibration certificate
- Non-roll end cap
- Supplied as standard in protective individual, resealable telescopic tube
- Made in UK
Specifications
Specifications
| Calibration Points | 0°C, 50°C, 100°C |
| Certifications/Compliance | ISO 17025 UKAS |
| Immersion Type | Partial Immersion |
| Length (Metric) | 40.5 cm |
| Material | Glass |
| Thermometer Fill | Blue Spirit |
| Max. Temperature (English) | 214°F |
| Max. Temperature (Metric) | +101°C |
| Min. Temperature (English) | 30°F |
| Min. Temperature (Metric) | -1°C |
| Temperature Range (English) | 30°F to 214°F |
| Temperature Range (Metric) | -1°C to 101°C |
| Accuracy | ± 1.0°C at 90°C |
| Subdivision | 0.2°C |
| Immersion Length | 76 mm |
| Width (Metric) | 1.2 cm |
| Show More Show Less |