Learn More
Lixivaptan, MedChemExpress
Product Code. 16483406
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483406 25 mg 25mg
16475988 10 mg 10mg
16468607 5 mg 5mg
16422137 50 mg 50mg
16410216 2 mg 2mg
16406416 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16483406 Supplier Medchem Express Supplier No. HY14185
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
MedChemExpress Lixivaptan (VPA-985, WAY-VPA 985) is an orally active and selective vasopressin receptor V2 antagonist, with IC50 values of 1.2 and 2.3 nM for human and rat V2, respectively.
Lixivaptan (VPA-985, WAY-VPA 985) is an orally active and selective vasopressin receptor V2 antagonist, with IC50 values of 1.2 and 2.3 nM for human and rat V2, respectively.
Specifications
Chemical Name or Material Lixivaptan
CAS 168079-32-1
Color White
Purity Grade Notes Research
Molecular Formula C27H21ClFN3O2
Quantity 25 mg
Synonym VPA-985 WAY-VPA 985
Solubility Information DMSO : ≥ 150 mg/mL (316.50 mM)
SMILES O=C(C1=C(Cl)C=C(NC(C2=CC(F)=CC=C2C)=O)C=C1)N(C3)C4=CC=CC=C4CN5C3=CC=C5
Molecular Weight (g/mol) 473.93
Formula Weight 473.93
Percent Purity 99.21%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation