missing translation for 'onlineSavingsMsg'
Learn More

Lithium Sulfate Monohydrate, Supelco™

Product Code. 15877380
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15877380 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ100 Graded - For Non-regulated Laboratory Applications
Product Code. 15877380 Supplier Supelco Supplier No. 1056970050
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ100 Graded - For Non-regulated Laboratory Applications

99.99 Suprapur™

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 120°C
CAS 10102-25-7
Quantity 50 g
MDL Number MFCD00149766
Synonym Lithium sulfate monohydrate, Lithium sulfate monohydrate
Molecular Weight (g/mol) 109.94 (Anhydrous Basis)
Grade Research
Density 2.05 g/cm3 at 20°C
Chemical Name or Material Lithium Sulfate Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.