Learn More
Lithium Carbonate, Supelco™
Description
We are committed to bringing you Greener Alternative Products, which belong to one of the four categories of greener alternatives. LITHIUM CARBONATE is essential for sustainable energy solutions, particularly in lithium-ion batteries. Its eco-friendly properties enhance battery performance and longevity, supporting the transition to cleaner energy technologies while reducing environmental impact.
Specifications
Specifications
| Melting Point | 720°C |
| CAS | 554-13-2 |
| Quantity | 250 g |
| MDL Number | MFCD00011084 |
| Synonym | Lithium carbonate, Lithium carbonate |
| Molecular Weight (g/mol) | 73.89 |
| Grade | Research |
| Density | 2.1 g/cm3 at 20°C |
| Chemical Name or Material | Lithium Carbonate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.