missing translation for 'onlineSavingsMsg'
Learn More

Lithium Acetate Dihydrate MP Biomedicals

Product Code. 11463452 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463452 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11463452 Supplier MP Biomedicals Supplier No. 0215525691
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6108-17-4
Quantity 1 kg
Molecular Formula C2H7LiO4
Synonym lithium acetate dihydrate, lithiumacetatedihydrate, lithium acetate dihydrate, reagent grade, lioac-2h2o, lioac.2h2o, ksc498a1d, c2h3o2.li.2h2o, lithotab acetate ion dihydrate, acetic acid lithium salt dihydrate
InChI Key IAQLJCYTGRMXMA-UHFFFAOYSA-M
SMILES [Li+].CC(=O)[O-].O.O
IUPAC Name lithium;acetate;dihydrate
Molecular Weight (g/mol) 102.014
PubChem CID 23666338
Formula Weight 102
Density 1.3 g/mL
Chemical Name or Material Lithium Acetate, Dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.