missing translation for 'onlineSavingsMsg'
Learn More

Lithium acetate dihydrate, MP Biomedicals™

Product Code. 11453452 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453452 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11453452 Supplier MP Biomedicals Supplier No. 0215525683
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Especificaciones

CAS 6108-17-4
Density 1.3g/mL
Molecular Formula C2H7LiO4
Quantity 250 g
Synonym lithium acetate dihydrate, lithiumacetatedihydrate, lithium acetate dihydrate, reagent grade, lioac-2h2o, lioac.2h2o, ksc498a1d, c2h3o2.li.2h2o, lithotab acetate ion dihydrate, acetic acid lithium salt dihydrate
InChI Key IAQLJCYTGRMXMA-UHFFFAOYSA-M
SMILES [Li+].CC(=O)[O-].O.O
IUPAC Name lithium;acetate;dihydrate
Molecular Weight (g/mol) 102.014
PubChem CID 23666338
Formula Weight 102
Physical Form Powder
Chemical Name or Material Lithium acetate dihydrate
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.