Learn More
Lisinopril dihydrate, MedChemExpress
Product Code. 16499957
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 16499957
Brand: Medchem Express HY18206A
MedChemExpress Lisinopri dihydrate (MK-521 dihydrate) is angiotensin-converting enzyme inhibitor, used in treatment of hypertension, congestive heart failure, and heart attacks.
Lisinopri dihydrate (MK-521 dihydrate) is angiotensin-converting enzyme inhibitor, used in treatment of hypertension, congestive heart failure, and heart attacks.
Specifications
Chemical Name or Material Lisinopril dihydrate
CAS 83915-83-7
Color White
Purity Grade Notes Research
Molecular Formula C21H35N3O7
Quantity 5 g
Synonym MK-521 dihydrate
Solubility Information H2O : 33.33 mg/mL (75.49 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (2.26 mM; Need ultrasonic)
SMILES O=C(O)[C@H]1N(C([C@H](CCCCN)N[C@H](C(O)=O)CCC2=CC=CC=C2)=O)CCC1.O.O
Molecular Weight (g/mol) 441.52
Formula Weight 441.52
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus