missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Liraglutide
GREENER_CHOICE

Product Code. 18757606
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18757606 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18757606 Supplier Tocris Bioscience™ Supplier No. 6517/1
  • ONLINE EXCLUSIVE PRICE
    565.00 EUR
    534.45 EUR / 1mg
    Save 30.55 EUR
    5% Off
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Highly potent, long-acting GLP-1 receptor agonist

Liraglutide is a highly potent, long-acting GLP-1 receptor agonist (EC50 = 61 pM). Acylated derivative of GLP-1 (7-37) (Cat. No. 5374). Inhibits food and water intake, causing lasting and reversible weight loss in normal and obese rats. In an animal model of Alzheimer's disease, Liraglutide decreases levels of Aβ and soluble amyloid, and reduces neuroinflammation.
TRUSTED_SUSTAINABILITY

Specifications

Purity 0.95
CAS 204656-20-2
Content And Storage Store at -20°C
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.