missing translation for 'onlineSavingsMsg'
Learn More

Liraglutide, TRC

Product Code. 30009829
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009829 25 mg 25mg
17973075 1 mg 1mg
17981575 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009829 Supplier Toronto Research Chemicals Supplier No. TRCL46886025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 204656-20-2
Molecular Formula C172H265N43O51
Molecular Weight (g/mol) 3751.2
Synonym L-Histidyl-L-alanyl-L-α-glutamylglycyl-L-threonyl-L-phenylalanyl-L-threonyl-L-seryl-L-α-aspartyl-L-valyl-L-seryl-L-seryl-L-tyrosyl-L-leucyl-L-α-glutamylglycyl-L-glutaminyl-L-alanyl-L-alanyl-N6-[N-(1-oxohexadecyl)-L-γ-glutamyl]-L-lysyl-L-α-glutamyl-L-phenylalanyl-L-isoleucyl-L-alanyl-L-tryptophyl-L-leucyl-L-valyl-L-arginylglycyl-L-arginylglycine
SMILES O=C(CCCCCCCCCCCCCCC)N[C@@H](CCC(NCCCC[C@@H](C(N[C@@H](CCC(O)=O)C(N[C@H](C(N[C@]([C@H](CC)C)([H])C(N[C@H](C(N[C@H](C(N[C@@H](CC(C)C)C(N[C@@H](C(C)C)C(N[C@@H](CCCNC(N)=N)C(NCC(N[C@@H](CCCNC(N)=N)C(NCC(O)=O)=O)=O)=O)=O)=O)=O)CC1=CNC2=C1C=CC=C2)=O)C)=O)=O)CC3=CC=CC=C3)=O)=O)NC([C@@H](NC([C@@H](NC([C@H](CCC(N)=O)NC(CNC([C@H](CCC(O)=O)NC([C@H](CC(C)C)NC([C@@H](NC([C@H](CO)NC([C@H](CO)NC([C@H](C(C)C)NC([C@@H](NC([C@H](CO)NC([C@@]([C@@H](C)O)([H])NC([C@@H](NC([C@@]([C@@H](C)O)([H])NC(CNC([C@H](CCC(O)=O)NC([C@@H](NC([C@@H](N[H])CC4=CN=CN4)=O)C)=O)=O)=O)=O)CC5=CC=CC=C5)=O)=O)=O)CC(O)=O)=O)=O)=O)=O)CC6=CC=C(O)C=C6)=O)=O)=O)=O)=O)C)=O)C)=O)=O)C(O)=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C172H265N43O51/c1-18-20-21-22-23-24-25-26-27-28-29-30-37-53-129(224)195-116(170(265)266)59-64-128(223)180-68-41-40-50-111(153(248)199-115(62-67-135(232)233)154(249)204-120(73-100-44-33-31-34-45-100)159(254)214-140(93(11)19-2)167(262)192-97(15)146(241)201-122(76-103-79-183-108-49-39-38-48-106(103)108)157(252)203-118(72-90(5)6)158(253)212-138(91(7)8)165(260)200-110(52-43-70-182-172(177)178)149(244)184-81-130(225)193-109(51-42-69-181-171(175)176)148(243)187-84-137(236)237)196-144(239)95(13)189-143(238)94(12)191-152(247)114(58-63-127(174)222)194-131(226)82-185-151(246)113(61-66-134(230)231)198-155(250)117(71-89(3)4)202-156(251)119(75-102-54-56-105(221)57-55-102)205-162(257)124(85-216)208-164(259)126(87-218)209-166(261)139(92(9)10)213-161(256)123(78-136(234)235)206-163(258)125(86-217)210-169(264)142(99(17)220)215-160(255)121(74-101-46-35-32-36-47-101)207-168(263)141(98(16)219)211-132(227)83-186-150(245)112(60-65-133(228)229)197-145(240)96(14)190-147(242)107(173)77-104-80-179-88-188-104/h31-36,38-39,44-49,54-57,79-80,88-99,107,109-126,138-142,183,216-221H,18-30,37,40-43,50-53,58-78,81-87,173H2,1-17H3,(H2,174,222)(H,179,188)(H,180,223)(H,184,244)(H,185,246)(H,186,245)(H,187,243)(H,189,238)(H,190,242)(H,191,247)(H,192,262)(H,193,225)(H,194,226)(H,195,224)(H,196,239)(H,197,240)(H,198,250)(H,199,248)(H,200,260)(H,201,241)(H,202,251)(H,203,252)(H,204,249)(H,205,257)(H,206,258)(H,207,263)(H,208,259)(H,209,261)(H,210,264)(H,211,227)(H,212,253)(H,213,256)(H,214,254)(H,215,255)(H,228,229)(H,230,231)(H,232,233)(H,234,235)(H,236,237)(H,265,266)(H4,175,176,181)(H4,177,178,182)/t93-,94-,95-,96-,97-,98+,99+,107-,109-,110-,111-,112-,113-,114-,115-,116-,117-,118-,119-,120-,121-,122-,123-,124-,125-,126-,138-,139-,140-,141-,142-/m0/s1
Formula Weight 3748.9465
Percent Purity >95%
Chemical Name or Material Liraglutide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.