Learn More
Labconco™ Liquid Trap Kit
Product Code. 15363476 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
CentriVap Concentrators and RapidVap Evaporators
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15363476 CentriVap Concentrators and RapidVap Evaporators Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15363476 Supplier Labconco™ Supplier No. 7970500
  • ONLINE EXCLUSIVE PRICE
    565.00 EUR
    536.85 EUR / Pack of 1
    Save 28.15 EUR
    4% Off
 Restricted Item
Estimated Shipment: 13-01-2027
For use with CentriVap™ Vacuum Concentrators and RapidVap™ Vacuum Evaporation Systems.
  • Liquid Trap prevents liquid from entering the pump
  • Includes borosilicate glass container that holds 240 milliliters of liquid, epoxy-coated steel bracket for attachment to the side of a CentriVap Concentrator or RapidVap Vacuum Evaporation System, 18 in. (457mm) of 0.5 in. ID (13mm) vacuum tubing, and two vacuum tubing clamps
Specifications
Type Liquid Trap
For Use With (Equipment) CentriVap Concentrators and RapidVap Evaporators