missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Linsitinib, MedChemExpress
Description
Linsitinib (OSI-906) is a potent, selective and orally bioavailable dual inhibitor of the IGF-1 receptor and insulin receptor (IR) with IC50s of 35 and 75 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | Linsitinib |
| CAS | 867160-71-2 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C26H23N5O |
| Quantity | 100 mg |
| Synonym | OSI-906 |
| Solubility Information | DMSO : 50 mg/mL (118.63 mM; Need ultrasonic) |
| SMILES | O[C@@]1(C)C[C@@H](C2=NC(C3=CC=C4C=CC(C5=CC=CC=C5)=NC4=C3)=C6C(N)=NC=CN62)C1 |
| Molecular Weight (g/mol) | 421.49 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?