missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Linsitinib

Description
Linsitinib is a potent and selective dual inhibitor of the IGF-1 receptor (IC50 = 35 nM) and insulin receptor (IC50 = 75 nM); it inhibits ligand-dependent auto-phosphorylation of both receptor tyrosine kinases and prevents ligand-induced activation of downstream pathways including pAKT, pERK1/2 and p-p70S6K. The compound is selective for IGF-1R, IR and IRR in a panel of 88 kinases. Lisintinib inhibits proliferation in a variety of tumor cell lines and shows robust in vivo antitumor efficacy in an IGF-1R-driven LSN xenograft model. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | cis-3-[8-Amino-1-(2-phenyl-7-quinolinyl)imidazo[1,5-a]pyrazin-3-yl]-1-methylcyclobutanol |
| CAS | 867160-71-2 |
| Quantity | 10 mg |
| Target | IGF-I R/IGF1R |
| Molecular Formula | C26H23N5O |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?