missing translation for 'onlineSavingsMsg'
Learn More

Limonin, >99.6%, For HPLC analysis, MP Biomedicals™

Product Code. 11482341 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 11482341

Brand: MP Biomedicals 0219376550

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Limonin from Citrus Seeds White Crystalline Powder

  • It may be used in assays to quantify limonin in citrus products.
  • It may also be used in other assays as a control, standard, or competitor or in studies on the physiology, metabolism, and biosynthesis of limonin.
  • It is a natural compound found in grapefruit and other citrus seeds.
  • IT is a triterpenoid aglycone that is a bitter principle of citrus fruits.
  • It has been shown to inhibit chemically induced carcinogenesis and demonstrates anti feedant properties.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1180-71-8
Molecular Formula C26H30O8
Molecular Weight (g/mol) 470.52
MDL Number MFCD00075922
InChI Key KBDSLGBFQAGHBE-MSGMIQHVSA-N
PubChem CID 179651
ChEBI CHEBI:16226
IUPAC Name (1R,2R,7S,10R,13R,14R,16S,19S,20S)-19-(furan-3-yl)-9,9,13,20-tetramethyl-4,8,15,18-tetraoxahexacyclo[11.9.0.0²,⁷.0²,¹⁰.0¹⁴,¹⁶.0¹⁴,²⁰]docosane-5,12,17-trione
SMILES CC1(C)O[C@H]2CC(=O)OC[C@@]22[C@H]1CC(=O)[C@]1(C)[C@@H]2CC[C@@]2(C)[C@@H](OC(=O)[C@H]3O[C@@]123)C1=COC=C1

Specifications

Color White
Assay Percent Range >99.6%
Quantity 50 mg
Synonym limonin, dictamnolactone, obaculactone, citrolimonin, evodin, 7,16-dioxo-7,16-dideoxylimondiol, limoni, limonoic acid, di-delta-lactone, limonine, limonoate d-ring-lactone
Formula Weight 470.5
Physical Form Powder
Chemical Name or Material Limonin, For HPLC analysis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.