missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lilial, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| CAS | 80-54-6 |
| Molecular Formula | C14H20O |
| MDL Number | MFCD00047655 |
| Quantity | 10 mg |
| Synonym | lilial, 3-4-tert-butyl phenyl-2-methylpropanal, 3-4-tert-butylphenyl isobutyraldehyde, lilyal, lily aldehyde, 3-4-tert-butylphenyl-2-methylpropanal, 3-4-tert-butylphenyl-2-methylpropionaldehyde, benzenepropanal, 4-1,1-dimethylethyl-.alpha.-methyl, butylphenyl methylpropional, 2-4-tert-butylbenzyl propionaldehyde |
| InChI Key | SDQFDHOLCGWZPU-UHFFFAOYNA-N |
| SMILES | CC(CC1=CC=C(C=C1)C(C)(C)C)C=O |
| IUPAC Name | 3-(4-tert-butylphenyl)-2-methylpropanal |
| Molecular Weight (g/mol) | 204.31 |
| PubChem CID | 228987 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction