Learn More
Thermo Scientific™ Light /UV Protection for Vertical Light Chambers
Product Code. 16461125 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
Factory Installed Door Switch
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
16461125 Factory Installed Door Switch Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16461125 Supplier Thermo Scientific™ Supplier No. 1900663
  • ONLINE EXCLUSIVE PRICE
    119.32 EUR
    108.00 EUR / Pack of 1
    Save 11.32 EUR
    9% Off
 Restricted Item
Estimated Shipment: 24-02-2027
Protection for operator, light switches off automatically when door is opened
Light / UV protection is a feature to protect the operator's eyes. This factory-installed option will switch off the vertical light as the door is opened. Recommended for ICH photostability testing. Not recommended for plant growth / animal hatching, sinc
  • Light / UV protection for operator's eyes.
  • Factory-installed option will switch off the vertical light as the door is opened
  • Recommended for ICH photo stability testing
  • Not recommended for plant growth/animal hatching since switching light on and off could stress plants and animals
  • Operator can wear goggles as an alternative protection.
TRUSTED_SUSTAINABILITY
Specifications
Description Light / UV protection - light off with open door
For Use With (Equipment) All Thermo Scientific and Forma vertical light chambers - models 3906, 3909, 3928, 3942, 3943
Lighting LED
Type Light/UV Protection
For Use With (Application) Light application - recommended for ICH photostability testing
Includes Factory Installed Door Switch