missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Imager UV/SYBR™ Filter

Product Code. 11632129
Change view
Click to view available options
For Use With (Equipment):
E-Gel Imager
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11632129 E-Gel Imager Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11632129 Supplier Invitrogen™ Supplier No. 4466608

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

1 x E-Gel™ Imager UV/SYBR™ Filter

Optimized filter for use with the E-Gel™ Imager system

Simply slide in this E-Gel Imager UV/SYBR™ Filter to replace the pre-installed universal filter for visualization of SYBR dyes like SYBR Safe when using UV transillumination rather than blue light transillumination.

  • Change filters easily without needing any tools
  • Get quality images quickly and easily using E-Gel Imager system
  • Filter Trays allow Easy Swapping with no extra tools
  • Each E-Gel Imager Camera Hood comes with a pre-installed universal filter
  • Alternative filters can be easily utilized by simply removing universal filter tray and sliding in filter tray of your choosing into camera hood
  • UV/SYBR Filter can be chosen when documenting gels stained with a SYBR stain using UV transillumination
  • Compact Solution for Agarose Gel Imaging, Analysis, and Documentation
  • Very powerful and flexible system that allows you to document gel images using multiple nucleic acid dyes
  • Each E-Gel Imager system includes a universal filter along with the camera hood

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 x E-Gel™ Imager UV/SYBR™ Filter.

Store at room temperature.
For Use With (Equipment) E-Gel Imager
Product Line E-Gel
Type Imager Filter
Filter Type Emission Filters
Label or Dye SYBR™ Safe
Shipping Condition Room Temperature

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.