Learn More
Lenz Laborglasintrumente™ Liebig Condenser with Olives
Product Code. 10200981
Change view
Click to view available options
Standard Taper Joint No:
NS 14/23
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Unit Size
10200981 NS 14/23 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10200981 Supplier Lenz Laborglasintrumente™ Supplier No. 05210201
  • ONLINE EXCLUSIVE PRICE
    53.00 EUR
    41.91 EUR / Pack of 1
    Save 11.09 EUR
    20% Off
Estimated Shipment: 19-01-2027
Liebig condenser for condensing vapours
  • Liebig condenser acc. to DIN 12576 and in addition to DIN
  • With NS socket and NS cone
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications
Certifications/Compliance DIN 12576
Description Cooling Surface - 60 cm²
Length (Metric) Jacket 160 mm
Quantity 1
Standard Taper Joint No NS 14/23
Material DURAN™ Tubing