Learn More
Gilac™ Lid with Handle for Round HACCP Food Contact Container
Product Code. 17998691
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
For Use With (Equipment):
50 L Round HACCP Food Contact Container
75 L Round HACCP Food Contact Container
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color For Use With (Equipment) Unit Size
17998691 Yellow 50 L Round HACCP Food Contact Container Each
17978951 White 50 L Round HACCP Food Contact Container Each
17948741 Blue 50 L Round HACCP Food Contact Container Each
17968661 Green 50 L Round HACCP Food Contact Container Each
17968741 Red 50 L Round HACCP Food Contact Container Each
17988831 White 75 L Round HACCP Food Contact Container Each
17958911 Yellow 75 L Round HACCP Food Contact Container Each
17938581 Blue 75 L Round HACCP Food Contact Container Each
17958581 Red 75 L Round HACCP Food Contact Container Each
17968651 Green 75 L Round HACCP Food Contact Container Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 17998691 Supplier Gilac™ Supplier No. G613323
  • ONLINE EXCLUSIVE PRICE
    22.98 EUR
    15.63 EUR / Pack of 1
    Save 7.35 EUR
    31% Off
Estimated Shipment: 29-12-2026
Lid with handle for round HACCP container.
  • Lid with handle for round HACCP container.
Specifications
Product Type HACCP Food Container Lid
Material Polyethylene
Color Yellow
For Use With (Equipment) 50 L Round HACCP Food Contact Container