Learn More
Levosulpiride, MedChemExpress
Product Code. 16418586
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16418586 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16418586 Supplier Medchem Express Supplier No. HYB1059
MedChemExpress Levosulpiride (RV-12309) is the (S)-enantiomer of sulpiride, which is a D2 receptor a antagonist, an atypical antipsychotic drug of the benzamide class.
Levosulpiride (RV-12309) is the (S)-enantiomer of sulpiride, which is a D2 receptor a antagonist, an atypical antipsychotic drug of the benzamide class.
Specifications
Chemical Name or Material Levosulpiride
CAS 23672-07-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H23N3O4S
Quantity 100 mg
Synonym RV-12309 S-(-)-Sulpiride
Solubility Information DMSO : 50 mg/mL (146.44 mM; Need ultrasonic)
SMILES O=C(NC[C@H]1N(CC)CCC1)C2=CC(S(=O)(N)=O)=CC=C2OC
Molecular Weight (g/mol) 341.43
Formula Weight 341.43
Percent Purity 99.87%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation