missing translation for 'onlineSavingsMsg'
Learn More

Levopride, TRC

Product Code. 17983265
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17983265 5 g 5g
17954095 100 mg 100mg
17942095 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17983265 Supplier Toronto Research Chemicals Supplier No. TRCL3760005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23672-07-3
Molecular Formula C15 H23 N3 O4 S
Molecular Weight (g/mol) 341.43
Synonym 5-(Aminosulfonyl)-N-[[(2S)-1-ethyl-2-pyrrolidinyl]methyl]-2-methoxybenzamide, Benzamide, 5-(aminosulfonyl)-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2-methoxy-, (S)-, o-Anisamide, N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-sulfamoyl-, (-)- (8CI), (-)-Sulpiride, L-Sulpiride, Lesuride, Levobren, Levopraid, Levosulpiride, S-(-)-Sulpiride, S-Sulpiride, l-Sulpiride, levopride, sulpid
IUPAC Name N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2-methoxy-5-sulfamoylbenzamide
SMILES CCN1CCC[C@H]1CNC(=O)c2cc(ccc2OC)S(=O)(=O)N

Specifications

Quantity 5 g
InChI Formula InChI=1S/C15H23N3O4S/c1-3-18-8-4-5-11(18)10-17-15(19)13-9-12(23(16,20)21)6-7-14(13)22-2/h6-7,9,11H,3-5,8,10H2,1-2H3,(H,17,19)(H2,16,20,21)/t11-/m0/s1
Formula Weight 341.1409
Percent Purity >95%
Chemical Name or Material Levosulpiride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.