missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Levoleucovorin Calcium, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
1g
2g
500mg
Description
Levoleucovorin calcium is the calcium salt of Levoleucovorin, which is the enantiomerically active form of folinic acid.
Specifications
Specifications
| Chemical Name or Material | Levoleucovorin Calcium |
| Physical Form | Solid |
| CAS | 80433-71-2 |
| Color | Light Yellow |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C20H21CaN7O7 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | Calcium levofolinate CL307782 |
| Solubility Information | H2O : 10 mg/mL (19.55 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction