Learn More
Levofloxacin-d3 Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Levofloxacin-d3 Sodium Salt |
| CAS | 2469039-38-9 |
| Molecular Formula | C18H16D3FN3NaO4 |
| Quantity | 1 mg |
| Synonym | (S)-9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic Acid-d3 Sodium Salt |
| InChI Formula | InChI=1S/C18H20FN3O4.Na/c1-10-9-26-17-14-11(16(23)12(18(24)25)8-22(10)14)7-13(19)15(17)21-5-3-20(2)4-6-21;/h7-8,10H,3-6,9H2,1-2H3,(H,24,25);/q;+1/p-1/t10-;/m0./s1/i2D3; |
| SMILES | FC1=CC2=C3C(OC[C@H](C)N3C=C(C(O[Na])=O)C2=O)=C1N4CCN(C([2H])([2H])[2H])CC4 |
| IUPAC Name | sodium (S)-9-fluoro-3-methyl-10-(4-(methyl-d3)piperazin-1-yl)-7-oxo-2,3-dihydro-7H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylate |
| Molecular Weight (g/mol) | 386.37 |
| Formula Weight | 386.14 |