Learn More
Levcromakalim, MedChemExpress
Product Code. 16462147
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16462147 50 mg 50mg
16436008 10 mg 10mg
16408627 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16462147 Supplier Medchem Express Supplier No. HY14255
MedChemExpress Levcromakalim ((-)-Cromakalim) is an ATP-sensitive K+ channel (KATP) activator.
Levcromakalim ((-)-Cromakalim) is an ATP-sensitive K+ channel (KATP) activator.
Specifications
Chemical Name or Material Levcromakalim
CAS 94535-50-9
Color White
Purity Grade Notes Research
Molecular Formula C16H18N2O3
Quantity 50 mg
Synonym (-)-Cromakalim BRL 38227
Solubility Information DMSO : ≥ 50 mg/mL (174.62 mM)
SMILES CC(O1)(C)[C@@H](O)[C@H](N2CCCC2=O)C3=C1C=CC(C#N)=C3
Molecular Weight (g/mol) 286.33
Formula Weight 286.33
Percent Purity 99.84%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation