missing translation for 'onlineSavingsMsg'
Learn More

Leu-AMS, MedChemExpress

Product Code. 16404248
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16404248 10 mg 10mg
16431119 1 mg 1mg
16416967 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16404248 Supplier Medchem Express Supplier No. HY108900
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Leu-AMS (compound 6), a leucine analogue, is a potent inhibitor of leucyl-tRNA synthetase (LRS) with an IC50 of 22.34 nM, which inhibits the catalytic activity of LRS but did not affect the leucine-induced mTORC1 activation. Leu-AMS shows cytotoxicity in cancer cells and normal cells, and inhibits the growth of bacteria.

Leu-AMS (compound 6), a leucine analogue, is a potent inhibitor of leucyl-tRNA synthetase (LRS) with an IC50 of 22.34 nM, which inhibits the catalytic activity of LRS but did not affect the leucine-induced mTORC1 activation. Leu-AMS shows cytotoxicity in cancer cells and normal cells, and inhibits the growth of bacteria.

Specifications

Chemical Name or Material Leu-AMS
CAS 288591-93-5
Color White
Purity Grade Notes Research
Molecular Formula C16H25N7O7S
Quantity 10 mg
Solubility Information DMSO : ≥ 49.17 mg/mL (107.01 mM)
SMILES O[C@@H]([C@H]([C@H](N1C=NC2=C1N=CN=C2N)O3)O)[C@H]3COS(=O)(NC([C@@H](N)CC(C)C)=O)=O
Molecular Weight (g/mol) 459.48
Formula Weight 459.48
Percent Purity 97.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.