missing translation for 'onlineSavingsMsg'
Learn More

Letrazuril, MedChemExpress

Product Code. 16486797
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486797 5 mg 5mg
16465066 100 mg 100mg
16454098 10 mg 10mg
16430457 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16486797 Supplier Medchem Express Supplier No. HY106859
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Letrazuril is an anti-HIV agent.

Letrazuril is an anti-HIV agent.

Specifications

Chemical Name or Material Letrazuril
CAS 103337-74-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H9Cl2FN4O2
Quantity 5 mg
Solubility Information DMSO : ≥ 165 mg/mL (421.80 mM)
SMILES O=C(N1)N(N=CC1=O)C2=CC(Cl)=C(C(C3=CC=C(F)C=C3)C#N)C(Cl)=C2
Molecular Weight (g/mol) 391.18
Formula Weight 391.18
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.