Learn More
Leonurine hydrochloride, MedChemExpress
Product Code. 16499078
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499078 10 mg 10mg
16484877 50 mg 50mg
16443896 25 mg 25mg
16439136 100 mg 100mg
16421498 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16499078 Supplier Medchem Express Supplier No. HYN0741A
MedChemExpress Leonurine hydrochloride is an alkaloid isolated from Herba leonuri, with anti-oxidative and anti-inflammatory.
Leonurine hydrochloride is an alkaloid isolated from Herba leonuri, with anti-oxidative and anti-inflammatory.
Specifications
Chemical Name or Material Leonurine hydrochloride
CAS 24735-18-0
Color White
Purity Grade Notes Research
Molecular Formula C14H22ClN3O5
Quantity 10 mg
Synonym SCM-198 hydrochloride
Solubility Information DMSO : ≥ 31 mg/mL (89.13 mM) ∣H2O : 5 mg/mL (14.38 mM; Need ultrasonic)
SMILES O=C(OCCCCNC(N)=N)C1=CC(OC)=C(O)C(OC)=C1.[H]Cl
Molecular Weight (g/mol) 347.79
Formula Weight 347.79
Percent Purity 99.69%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation