missing translation for 'onlineSavingsMsg'
Learn More

Lenvatinib, MedChemExpress

Product Code. 16464328
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464328 10 mg 10mg
16445246 100 mg 100mg
16437047 5 mg 5mg
16420657 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16464328 Supplier Medchem Express Supplier No. HY10981
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Lenvatinib (E7080) is an oral, multi-targeted tyrosine kinase inhibitor that inhibits VEGFR1-3, FGFR1-4, PDGFR, KIT, and RET, shows potent antitumor activities.

Lenvatinib (E7080) is an oral, multi-targeted tyrosine kinase inhibitor that inhibits VEGFR1-3, FGFR1-4, PDGFR, KIT, and RET, shows potent antitumor activities.

Specifications

Chemical Name or Material Lenvatinib
CAS 417716-92-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H19ClN4O4
Quantity 10 mg
Synonym E7080
Solubility Information DMSO : 12.78 mg/mL (29.94 mM; Need ultrasonic)
SMILES O=C(N)C1=C(C=C2N=CC=C(C2=C1)OC3=CC=C(C(Cl)=C3)NC(NC4CC4)=O)OC
Molecular Weight (g/mol) 426.85
Formula Weight 426.85
Percent Purity 99.22%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.