missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Lenalidomide 5'-piperazine
GREENER_CHOICE

Product Code. 18737546
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 18737546

Brand: Tocris Bioscience™ 7472/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cereblon ligand with terminal piperazine for onward chemistry

Lenalidomide 5'-piperazine is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand with a terminal piperazine for onward chemistry to generate Protein Degraders with rigid linkers. Part of a range of functionalized tool molecules for PROTAC R&D.

PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[1,3-Dihydro-1-oxo-5-(1-piperazinyl)-2 H-isoindol-2-yl]-2,6-piperidinedione hydrochloride
CAS 2222120-31-0
Quantity 25 mg
Target Degrader Building Blocks
Molecular Formula C17H20N4O3.HCl
Purity 0.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.