Learn More
Gibco™ Leibovitz's L-15 Medium, GlutaMAX™ Supplement
GREENER_CHOICE
Product Code. 11544436
Change view
Click to view available options
Quantity:
500 mL
10 x 500 mL
Unit Size:
500mL
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11544436 10 x 500 mL Pack of 10
11534436 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11544436 Supplier Gibco™ Supplier No. 31415086
Edge
Leibovitz's L-15 Medium supports the growth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue. L-15 is buffered by phosphates and free base amino acids instead of sodium bicarbonate.
Leibovitz's L-15 Medium supports the growth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue. L-15 is buffered by phosphates and free base amino acids instead of sodium bicarbonate. This medium is designed for supporting cell growth in environments without CO2 equilibration.
This L-15 is modified as follows:
With: Galactose, Phenol Red, GlutaMAX™, Sodium Pyruvate
Without: Glucose, HEPES, Sodium Bicarbonate
The complete formulation is available.
Gibco™ L-15 with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.
L-15 contains no proteins, lipids, or growth factors. Therefore, L-15 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS).
TRUSTED_SUSTAINABILITY
Specifications
Concentration 1 X
Form Liquid
Product Type Leibovitz's L-15 Medium
With Additives Sodium Pyruvate
Without Additives No Glucose, No HEPES, No Sodium Bicarbonate
For Use With (Application) Mammalian cell culture
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Quantity 10 x 500 mL
Certifications/Compliance ISO 13485 standards, FDA
pH 7.4 to 7.9
Regulatory Status For in-vitro diagnostic use
Content And Storage 2 to 8°C
Description Leibovitz's L-15 Medium, 500mL, 1 X Concentration, Liquid Form, With Galactose, Phenol Red, GlutaMAX™, Sodium Pyruvate, 7.4 to 7.9 pH, 12 Months Shelf Life, 300 to 340mOsm/kg Osmolality, Used for Mammalian Cell Culture
Show More Show Less
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.