Learn More
Thermo Scientific™ Legionella GVPC Selective Agar
Click to view available options
Quantity:
10/Pk.
Format:
90 mm Monoplate
Media Type:
Legionella GVPC Selective Agar
Manufacturing Origin:
Germany
Unit Size:
Pack of 10
Description
Isolate Legionella from clinical specimens and environmental samples using Thermo Scientific™ Legionella GVPC Selective Agar, a selective medium that conforms to ISO and exhibits optimal recovery of Legionellaceae in short incubation periods from both environmental samples and clinical specimens.
Legionella are aerobic, motile, nutritionally fastidious, pleomorphic, gram-negative rods that require special growth factors in culture media to grow. Transmission occurs through aerosolization or aspiration of contaminated water, not person to person. Legionellosis refers to two distinct clinical syndromes, Legionnaires’ disease, which most often presents as severe pneumonia, and Pontiac fever, which is an acute, febrile, self-limiting, viral-like illness. Over forty species of Legionella have been characterized, however Legionella pneumophila remains the most prevalent etiological agent of Legionnaires’ disease. Legionella can be isolated from respiratory tract secretions of all types, sterile body fluids such as blood, and biopsy material.
This product may not be available in all countries or territories. Please consult your local representative for availability.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
This product may not be available in all countries or territories. Please consult your local representative for availability.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
Specifications
Specifications
| Certifications/Compliance | According to ISO 11731-2 |
| Color | Traffic Black to Jet black, opaque |
| Content And Storage | 2° to 12°C, in the dark |
| Format | 90 mm Monoplate |
| Manufacturing Origin | Germany |
| Media Type | Legionella GVPC Selective Agar |
| Product Type | Agar |
| Fill Weight | 20 g ± 5 % |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 10/Pk. |