Learn More
Novacyt - Microgen Bioproducts Legionella 2-15 Latex Reagent
Product Code. 17313201 Shop All Novacyt Group Products
Change view
Click to view available options
Quantity:
2.5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17313201 2.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17313201 Supplier Novacyt - Microgen Bioproducts Supplier No. M45B
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Microgen Bioproducts Legionella 2-15 latex reagent
  • Individual L.pneumophila serogroups 1, 2-15, and non L.pneumophila species latex and positive control available separately.
Specifications
Certifications/Compliance ISO13485 ISO9001
Content And Storage 2°C to 8°C
CE Marker Yes
Clia Complexity Non-waived
Detectable Analytes L. pneumophila Serogroup 2-15 Antibodies
Format 1 Bottle (2.5 mL)
Sample Type Bacterial Colonies
Sensitivity 1
Type Reagent
For Use With (Application) The Confirmatory Identification of Legionella Pneumophila and Other Commonly Isolated Legionellasp Ecies Grown on Selective Medi
For Use With (Equipment) 0.85% Isotonic Saline, Disposable Agglutination Slides, Disposable Mixing Sticks, Bacteriological Loops, Legionella Selective Medium, Glass Tubes for Boiling
Includes 2.5 mL
Material Polyethylene Dropper Bottle
No. of Reactions 1/Test
Organism Group Monera
Packaging Type Bottle in polygrip bag
Quantity 2.5 mL
Reagent Type M45b Legionella Test Reagent 2-15 2.5 mL
Species Legionella pneumophila
Specificity 1
Target Organism Class Bacteria
Test Time 7 minutes