Learn More
Leflunomide, Thermo Scientific™
151.00 EUR - 455.00 EUR
Chemical Identifiers
| CAS | 75706-12-6 |
|---|---|
| Molecular Formula | C12H9F3N2O2 |
| Molecular Weight (g/mol) | 270.211 |
| MDL Number | MFCD00867593 |
| InChI Key | VHOGYURTWQBHIL-UHFFFAOYSA-N |
| Synonym | leflunomide, arava, leflunomidum, lefunamide, leflunomida, leflunomidum inn-latin, 5-methyl-n-4-trifluoromethyl phenyl-1,2-oxazole-4-carboxamide, leflunomida inn-spanish, su 101 pharmaceutical, arava tn |
| PubChem CID | 3899 |
| ChEBI | CHEBI:6402 |
| IUPAC Name | 5-methyl-N-[4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide |
| SMILES | CC1=C(C=NO1)C(=O)NC2=CC=C(C=C2)C(F)(F)F |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15447129
|
Thermo Scientific Alfa Aesar
J65917.MC |
100mg |
151.00 EUR
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15457129
|
Thermo Scientific Alfa Aesar
J65917.ME |
500mg |
455.00 EUR
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- An immunosuppressive agent
- Inhibits T and B cell proliferation
Chemical Identifiers
| 75706-12-6 | |
| 270.211 | |
| VHOGYURTWQBHIL-UHFFFAOYSA-N | |
| 3899 | |
| 5-methyl-N-[4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide |
| C12H9F3N2O2 | |
| MFCD00867593 | |
| leflunomide, arava, leflunomidum, lefunamide, leflunomida, leflunomidum inn-latin, 5-methyl-n-4-trifluoromethyl phenyl-1,2-oxazole-4-carboxamide, leflunomida inn-spanish, su 101 pharmaceutical, arava tn | |
| CHEBI:6402 | |
| CC1=C(C=NO1)C(=O)NC2=CC=C(C=C2)C(F)(F)F |
Specifications
| 75706-12-6 | |
| MFCD00867593 | |
| 14,5432 | |
| VHOGYURTWQBHIL-UHFFFAOYSA-N | |
| 5-methyl-N-[4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide | |
| 3899 | |
| 270.21 |
| C12H9F3N2O2 | |
| UN2811 | |
| leflunomide, arava, leflunomidum, lefunamide, leflunomida, leflunomidum inn-latin, 5-methyl-n-4-trifluoromethyl phenyl-1,2-oxazole-4-carboxamide, leflunomida inn-spanish, su 101 pharmaceutical, arava tn | |
| CC1=C(C=NO1)C(=O)NC2=CC=C(C=C2)C(F)(F)F | |
| 270.211 | |
| CHEBI:6402 | |
| Leflunomide |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'rtecsNumber' : NY2354200
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.