Learn More
Leflunomide, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J65917.MC
| Quantity | 100mg |
|---|
View more versions of this product
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- An immunosuppressive agent
- Inhibits T and B cell proliferation
Chemical Identifiers
| 75706-12-6 | |
| 270.211 | |
| VHOGYURTWQBHIL-UHFFFAOYSA-N | |
| 3899 | |
| 5-methyl-N-[4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide |
| C12H9F3N2O2 | |
| MFCD00867593 | |
| leflunomide, arava, leflunomidum, lefunamide, leflunomida, leflunomidum inn-latin, 5-methyl-n-4-trifluoromethyl phenyl-1,2-oxazole-4-carboxamide, leflunomida inn-spanish, su 101 pharmaceutical, arava tn | |
| CHEBI:6402 | |
| CC1=C(C=NO1)C(=O)NC2=CC=C(C=C2)C(F)(F)F |
Specifications
| 75706-12-6 | |
| MFCD00867593 | |
| UN2811 | |
| leflunomide, arava, leflunomidum, lefunamide, leflunomida, leflunomidum inn-latin, 5-methyl-n-4-trifluoromethyl phenyl-1,2-oxazole-4-carboxamide, leflunomida inn-spanish, su 101 pharmaceutical, arava tn | |
| CC1=C(C=NO1)C(=O)NC2=CC=C(C=C2)C(F)(F)F | |
| 270.211 | |
| CHEBI:6402 | |
| Leflunomide |
| C12H9F3N2O2 | |
| 100mg | |
| 14,5432 | |
| VHOGYURTWQBHIL-UHFFFAOYSA-N | |
| 5-methyl-N-[4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide | |
| 3899 | |
| 270.21 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.