missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leflunomide, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Leflunomide |
| CAS | 75706-12-6 |
| Analyte or Component Names | Leflunomide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H9 F3 N2 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Leflunomide, 5-Methyl-N-[4-(trifluoromethyl)phenyl]-4-isoxazolecarboxamide, Arava, HWA 486, Leflunomida, Leflunomidum, SU 101 |
| InChI Formula | InChI=1S/C12H9F3N2O2/c1-7-10(6-16-19-7)11(18)17-9-4-2-8(3-5-9)12(13,14)15/h2-6H,1H3,(H,17,18) |
| Solution Type | Neat |
| SMILES | Cc1oncc1C(=O)Nc2ccc(cc2)C(F)(F)F |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction