missing translation for 'onlineSavingsMsg'
Learn More

Ledipasvir acetone, Thermo Scientific™

Product Code. 17750709 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17750709 100 mg 100mg
17760709 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17750709 Supplier Thermo Scientific Acros Supplier No. 469361000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Chemical Name or Material Ledipasvir acetone
CAS 1441674-54-9
Quantity 100 mg
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C49 H54 F2 N8 O6 . C3 H6 O
Physical Form Powder
Color White to Yellow
Purity ≥97.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.